Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atrial Natriuretic peptide Rabbit anti-Mouse, Rat, Polyclonal, Chemicon™

Catalog No. AB5490I25UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5490-I-25UL 25 μL
AB5490-I-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB5490-I-25UL Supplier MilliporeSigma Supplier No. AB5490I25UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Atrial Natriuretic peptide is a highly specific rabbit polyclonal antibody, that targets Natriuretic peptides A and has been tested in Western Blotting and Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Atrial Natriuretic peptide
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Nppa
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 28 amino acids from the C-terminal region of rat Natriuretic peptides A.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_006163.1
Test Specificity This rabbit polyclonal antibody detects Natriuretic peptides A in rat and murine heart. It targets an epitope within 28 amino acids from the C-terminal region.
Target Species Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.