Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATRIP Antibody (OTI5E7), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272271PEATT594
Description
ATRIP Monoclonal specifically detects ATRIP in Human, Canine samples. It is validated for Flow Cytometry.Specifications
ATRIP | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
ATRIP | |
Recombinant protein fragment corresponding to amino acids 42-309 of human ATRIP (NP_569055) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Canine | |
IgG2a |
Flow Cytometry | |
OTI5E7 | |
Flow Cytometry | |
AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740 | |
Mouse | |
Affinity Purified | |
Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
84126.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction