Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATRIP Antibody (SR2262), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32239450UL
Description
ATRIP Monoclonal antibody specifically detects ATRIP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ATRIP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
SR2262 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740 | |
A synthesized peptide derived from human ATRIP (Uniprot #: Q8WXE1) | |
50 μg | |
Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
84126 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction