Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATRIP Monoclonal Antibody (OTI4A8), TrueMAB™, OriGene
SDP

Catalog No. 502540199
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0199 30 μL
50-167-5287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0199 Supplier OriGene Supplier No. TA504640S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATRIP Monoclonal antibody specifically detects ATRIP in Canine, Human, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen ATRIP
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CYB5R1
Gene Accession No. Q5EB81, Q9UHQ9
Gene Alias 6620401K05Rik; AGS1; ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR interacting protein; ATR IP; ATR-interacting protein; Atrip; DNase III; DRN3; FLJ12343; unnamed protein product
Host Species Mouse
Immunogen Full length human recombinant protein of human CYB5R1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304805, 51706, 606823
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.