Learn More
Invitrogen™ ATRIP Monoclonal Antibody (OTI5E7)

Description
ATRIP Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | ATRIP |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI5E7 |
| Concentration | 1.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ATRIP |
| Gene Accession No. | Q8WXE1 |
| Gene Alias | 6620401K05Rik; AGS1; ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR interacting protein; ATR IP; ATR-interacting protein; Atrip; DNase III; DRN3; FLJ12343; unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.