Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATRIP Recombinant Rabbit Monoclonal Antibody (2C4B6)
GREENER_CHOICE

Catalog No. PIMA537906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537906 Supplier Invitrogen™ Supplier No. MA537906
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: MCF7, Jurkat, Mouse testis, Mouse brain, Rat testis, Rat brain Immunogen sequence: NELRTKLQTS ERANKLAAPS VSHVSPRKNP SVVIKPEACS PQFGKTSFPT KESFSANMSL PHPCQTESGY KPLVGREDSK PHSLRGDSIK QEEAQKSFVD S.

ATRIP is an 86 kDa protein that is phosphorylated by ATR, regulates ATR expression, and is an essential component of the DNA damage checkpoint pathway. It localizes to the intranuclear foci after DNA damage or inhibition of replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATRIP
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2C4B6
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATRIP
Gene Accession No. Q8BMG1, Q8WXE1
Gene Alias 6620401K05Rik; AGS1; ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR interacting protein; ATR IP; ATR-interacting protein; Atrip; DNase III; DRN3; FLJ12343; unnamed protein product
Gene Symbols ATRIP
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ATRIP (Q8WXE1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235610, 301014, 84126
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.