Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATRNL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981424
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1424 Supplier Bioss Supplier No. BS11504R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATRNL1 (Attractin Like 1) is a Protein Coding gene. May play a role in melanocortin signaling pathways that regulate energy homeostasis. Gene Ontology (GO) annotations related to this gene include carbohydrate binding.

Specifications

Antigen ATRNL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATRNL1
Gene Accession No. Q5VV63
Gene Alias AI504415; Alp; Atrnl; Atrnl1; Atrnlp; attractin like 1; attractin-like 1; Attractin-like protein 1; AW555641; bA338L11.1; bA454H24.1; Kiaa0534
Gene Symbols ATRNL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATRNL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26033
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.