Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atrophin 1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501558369
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8369 Supplier Bethyl Laboratories, Inc Supplier No. A300753A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Chimpanzee

Dentatorubral-pallidoluysian atrophy protein, also designated atrophin-1, interacts with several other proteins, including RERE, BAIAP2 and WWP1-3. It is highly expressed in ovary, testis, brain and prostate, but can also be detected in thymus, liver and leukocytes. Defects in ATN1, the gene encoding for the atrophin protein, can cause dentatorubral-pallidoluysian atrophy (DRPLA) or Haw River syndrome (HRS). Both disorders are domit neurodegenerative disorders caused by an increase in the number of polyglutamine (Gln) repeats in the ATN1 gene (7-23 repeats in the normal population, 49-75 in patients affected by DRPLA or HRS). More repeats corresponds to earlier onset and more severe clinical manifestations of the diseases. DRPLA is characterized by a loss of neurons in the dentate nucleus, rubrum, globus pallidus and Luys′ body, often resulting in dementia, epilepsy and cerebellar ataxia. HRS is characterized by the degeneration of multiple systems, resembling that of DRPLA or Huntington′s disease.

Specifications

Antigen Atrophin 1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ATN1
Gene Accession No. P54259
Gene Alias Atrophin1, Atrophin 1, Dentatorubral-pallidoluysian atrophy protein, ATN1, D12S755E, DRPLA
Gene Symbols ATN1
Host Species Rabbit
Immunogen Between 1150 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1822
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.