Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATRX Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993091
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3091 10 μL
50-199-3090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3091 Supplier Active Motif Supplier No. 61050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATRX encodes a protein that contains an ATPase/helicase domain, and thus it belongs to the SWI/SNF family of chromatin remodeling proteins. The mutations of this gene are associated with an X-linked mental retardation (XLMR) syndrome most often accompanied by alpha-thalassemia (ATRX) syndrome. These mutations have been shown to cause diverse changes in the pattern of DNA methylation, which may provide a link between chromatin remodeling, DNA methylation, and gene expression in developmental processes. This protein is found to undergo cell cycle-dependent phosphorylation, which regulates its nuclear matrix and chromatin association, and suggests its involvement in the gene regulation at interphase and chromosomal segregation in mitosis. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.

Specifications

Antigen ATRX
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene ATRX
Gene Accession No. P46100
Gene Alias 4833408C14Rik; AI447451; alpha thalassemia/mental retardation syndrome (X-linked); alpha thalassemia/mental retardation syndrome X-linked; alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S. cerevisiae); alpha thalassemia/mental retardation syndrome X-linked homolog; alpha thalassemia/mental retardation syndrome X-linked homolog (human); ATP-dependent helicase ATRX; ATR2; Atrx; ATRX, chromatin remodeler; DNA dependent ATPase and helicase; DXHXS6677E; helicase 2; helicase 2, X-linked; helicase II; Heterochromatin protein 2; HP1 alpha-interacting protein; Hp1bp2; Hp1bp38; HP1-BP38; HP1-BP38 protein; JMS; mental retardation, X-linked 52; MRX52; MRXHF1; MRXS3; pABP-2; RAD54; RAD54L; RP5-875J14.1; SFM1; SHS; Transcriptional regulator ATRX; XH2; X-linked helicase II; X-linked nuclear protein; Xnp; Zinc finger helicase; Znf-HX
Gene Symbols ATRX
Host Species Rabbit
Immunogen This ATRX antibody was raised against a peptide within the C-terminus of human ATRX.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 546
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.