Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega AttoPhos™ Buffer

The AttoPhos(R) Buffer is optimized for sensitive detection of alkaline phosphatase (AP) when used to prepare working solutions with AttoPhos(R) Substrate.

$132.00 - $285.00

Specifications

pH 10
Storage Requirements 4°C
View More Specs

Products 2
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
PRS1022
View Documents
Promega
S1022
240mL
Each for $285.00
Only null left
Add to Cart
 
PRS1021
View Documents
Promega
S1021
60mL
Each for $132.00
Only null left
Add to Cart
 
Description

Description

  • Produces a low fluorescence signal until enzymatically acted upon, yielding detection of AP to 0.1 attomole
  • Provides low background
  • Offers linear kinetics over five orders of magnitude of AP concentration
  • Has an unusually large Stokes' shift (≈120nm), which leads to higher detection sensitivity

Specifications

Specifications

10
4°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.