Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Attractin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532668
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532668 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532668 Supplier Invitrogen™ Supplier No. PA532668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Marmoset, Mouse, Rat, Dog, Bat, Hamster, Panda, Pig (100%); Gorilla, Monkey, Rabbit (94%); Elephant (88%); Bovine, Stickleback (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Multiple transcript variants encoding different isoforms exist for this gene. One of the isoforms is a membrane-bound protein with sequence similarity to the mouse mahogany protein, a receptor involved in controlling obesity. The other isoform is a secreted protein involved in the initial immune cell clustering during inflammatory responses that may regulate the chemotactic activity of chemokines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Attractin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ATRN
Gene Accession No. O75882, Q99J86, Q9WU60
Gene Alias ATRN; atrn {ECO:0000312; Attractin; attractin-2; AW558010; DPPT-L; KIAA0548; mahogany; mahogany homolog; mahogany protein; membrane attractin; mg; MGC126754; Mgca; mKIAA0548; Protein mahogany; protein zitter; RGD:69063}
Gene Symbols ATRN
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human ATRN / Attractin.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100101470, 100774269, 11990, 477170, 83526, 8455
Target Species Human, Mouse, Rat, Canine, Hamster, Pig, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.