Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Attractin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110754 Supplier Invitrogen™ Supplier No. PA5110754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLLSDILVFT SEQCDAHRSE AACLAAGPGI RCVWNTGSSQ CISWALATDE QEEKLKSECF SKRTLDHDRC DQHTDCYSCT ANTNDCHWCN DHCVPRNHSC SEGQISIFRY ENCPKDNPMY YCNKKTSCRS CALDQNCQWE Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 89%.

Multiple transcript variants encoding different isoforms exist for this gene. One of the isoforms is a membrane-bound protein with sequence similarity to the mouse mahogany protein, a receptor involved in controlling obesity. The other isoform is a secreted protein involved in the initial immune cell clustering during inflammatory responses that may regulate the chemotactic activity of chemokines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Attractin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATRN
Gene Accession No. O75882
Gene Alias ATRN; atrn {ECO:0000312; Attractin; attractin-2; AW558010; DPPT-L; KIAA0548; mahogany; mahogany homolog; mahogany protein; membrane attractin; mg; MGC126754; Mgca; mKIAA0548; Protein mahogany; protein zitter; RGD:69063}
Gene Symbols ATRN
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Attractin. Recombinant protein control fragment (Product #RP-90009).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.