Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Attribute Blocks
SCI_ED

Catalog No. s79408
Click to view available options
Product Type:
Attribute Block Set
Giant set in Plastic storage tray

Order Info

Desk set sizes range from 10cm2 to 30cm2. Giant set sizes range from 25cm2 to 75cm2.

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
Format Hands-on Learning
Product Type Attribute Block Set
Includes Five shapes, two sizes, two thicknesses and three color, convenient storage tray with shape sorter
For Use With (Application) Teaches shapes, sorting, patterns, size and counting
Subject Mathematics
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.