Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atxn1 Mouse anti-Human, Mouse, HRP, Clone:S76-8, Abnova™

Catalog No. 89333751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-751 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-751 Supplier Abnova Corporation Supplier No. MAB17080
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of mouse Atxn1.

Specifications

Antigen Atxn1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S76-8
Conjugate HRP
Description ataxin 1
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Immunoprecipitation
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Atxn1
Gene Alias Ataxin-1, Atx1, C85907, Sca1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 164-197 of mouse Atxn1.
Quantity 100 μg
Gene ID (Entrez) 20238
Target Species Human, Mouse
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.