Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATXN10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598972 Supplier Thermo Scientific Supplier No. PA598972
Add to Cart
Add to Cart

ATXN10 Polyclonal Antibody

ATXN10 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATXN10
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATXN10
Gene Accession No. Q9UBB4
Gene Alias AI325283; ataxin 10; ataxin 10 homolog; ataxin-10; ATXN10; Brain protein E46; brain protein E46 homolog; C77170; E46; E46L; HUMEEP; Neuronal beta-catenin-like protein; Sca10; spinocerebellar ataxia 10; spinocerebellar ataxia 10 homolog; spinocerebellar ataxia type 10 protein; Spinocerebellar ataxia type 10 protein homolog; TEG-169; testis expressed gene 169; Tex169
Gene Symbols ATXN10
Host Species Rabbit
Immunogen Recombinant Human Ataxin-10 protein (346-475AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25814
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.