Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATXN10/SCA10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981684
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1684 Supplier Bioss Supplier No. BS11806R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The autosomal dominant cerebellar ataxias (ADCAs) are a clinically and genetically heterogeneous group of disorders characterized by ataxia, dysarthria, dysmetria, and intention tremor. All ADCAs involve some degree of cerebellar dysfunction and a varying degree of signs from other components of the nervous system. A commonly accepted clinical classification (Harding, 1993) divides ADCAs into 3 different groups based on the presence or absence of associated symptoms such as brainstem signs or retinopathy. The presence of pyramidal and extrapyramidal symptoms and ophthalmoplegia makes the diagnosis of ADCA I, the presence of retinopathy points to ADCA II, and the absence of associated signs to ADCA III. Genetic linkage and molecular analyses revealed that ADCAs are genetically heterogeneous even within the various subtypes.

Specifications

Antigen ATXN10/SCA10
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATXN10
Gene Accession No. Q9UBB4
Gene Alias AI325283; ataxin 10; ataxin 10 homolog; Ataxin-10; Atxn10; Brain protein E46; Brain protein E46 homolog; C77170; E46; E46L; HUMEEP; Neuronal beta-catenin-like protein; Sca10; spinocerebellar ataxia 10; spinocerebellar ataxia 10 homolog; Spinocerebellar ataxia type 10 protein; Spinocerebellar ataxia type 10 protein homolog; TEG-169; testis expressed gene 169; Tex169
Gene Symbols ATXN10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATXN10/SCA10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25814
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.