Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATXN1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549084
Description
Predicted to react with mouse based on sequence homology.
Chromatin-binding factor that repress Notch signaling in the absence of Notch intracellular domain by acting as a CBF1 corepressor. Binds to the HEY promoter and might assist, along with NCOR2, RBPJ-mediated repression (PubMed:21475249). Can suppress ATXN1 cytotoxicity in spinocerebellar ataxia type 1 (SCA1). In concert with CIC and ATXN1, involved in brain development. [UniProt].
Specifications
ATXN1L | |
Polyclonal | |
Unconjugated | |
ATXN1L | |
ankyrin repeat and SOCS box containing 16; ankyrin repeat and SOCS box protein 16; ankyrin repeat and SOCS box-containing 16; ankyrin repeat and SOCS box-containing protein 16; ASB16; ASB-16; ataxin 1 like; ataxin 1-like; ataxin-1-like; ATXN1L; BOAT; BOAT1; Brother of ataxin-1; brother of ATXN1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
342371 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P0C7T5 | |
ATXN1L | |
KLH conjugated synthetic peptide between 385-411 amino acids from the Central region of human ATXN1L. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction