Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATXN1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566506
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566506 Supplier Invitrogen™ Supplier No. PA566506
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHSFNKAPSAT SPSGQLPHHS STQPLDLAPG RMPIYYQMSR LPAGYTLHET PPAGASPVLT PQESQS Highest antigen sequence identity to the following orthologs - mouse 87%, rat 84%.

Chromatin-binding factor that repress Notch signaling in the absence of Notch intracellular domain by acting as a CBF1 corepressor. Binds to the HEY promoter and might assist, along with NCOR2, RBPJ-mediated repression (PubMed:21475249). Can suppress ATXN1 cytotoxicity in spinocerebellar ataxia type 1 (SCA1). In concert with CIC and ATXN1, involved in brain development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATXN1L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATXN1L
Gene Accession No. P0C7T5
Gene Alias ankyrin repeat and SOCS box containing 16; ankyrin repeat and SOCS box protein 16; ankyrin repeat and SOCS box-containing 16; ankyrin repeat and SOCS box-containing protein 16; ASB16; ASB-16; ataxin 1 like; ataxin 1-like; ataxin-1-like; ATXN1L; BOAT; BOAT1; Brother of ataxin-1; brother of ATXN1
Gene Symbols ATXN1L
Host Species Rabbit
Immunogen Recombinant Human ATXN1L. Recombinant protein control fragment (Product #RP-107176).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342371
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.