Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATXN2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111686 Supplier Invitrogen™ Supplier No. PA5111686
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSYIPYNPQQ FPGQPAMMQP MAHYPSQPVF APMLQSNPRM LTSGSHPQAI VSSSTPQYPS AEQPTPQALY ATVHQSYPHH ATQLHAHQP Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 99%.

This gene encodes an ataxin type 2 related protein of unknown function. This protein is a member of the spinocerebellar ataxia (SCAs) family, which is associated with a complex group of neurodegenerative disorders. Several alternatively spliced transcripts encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATXN2L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATXN2L
Gene Accession No. Q8WWM7
Gene Alias A2D; A2LG; A2LP; A2RP; ataxin 2 like; ataxin 2 related protein; ataxin 2-like; ataxin-2 domain protein; Ataxin-2-like protein; Ataxin-2-related protein; ATXN2L; Atxnl2
Gene Symbols ATXN2L
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ATXN2L. Recombinant protein control fragment (Product #RP-105552).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11273
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.