Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATXN7L1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297598F
Description
ATXN7L1 Polyclonal specifically detects ATXN7L1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATXN7L1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ataxin 7-like 1, ataxin 7-like 4, ataxin-7-like protein 1, Ataxin-7-like protein 4, ATXN7L4, FLJ40255, FLJ58242, KIAA1218MGC10760, MGC33190 | |
Produced in rabbits immunized with E. coli-derived Human ATXN7L1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
222255 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction