Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATXN7L1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP29925150UL
Description
ATXN7L1 Polyclonal antibody specifically detects ATXN7L1 in Human samples. It is validated for Western BlotSpecifications
ATXN7L1 | |
Polyclonal | |
Western Blot 1:500-1:1000 | |
ataxin 7-like 1, ataxin 7-like 4, ataxin-7-like protein 1, Ataxin-7-like protein 4, ATXN7L4, FLJ40255, FLJ58242, KIAA1218MGC10760, MGC33190 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ATXN7L1. | |
50 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
222255 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction