Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATXN7L1 Monoclonal Antibody (OTI4F6)
GREENER_CHOICE

Catalog No. PIMA526196
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526196 Supplier Invitrogen™ Supplier No. MA526196
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATXN7L1 Monoclonal Antibody for Western Blot, IHC (P)

ATXN7L1 (ataxin-7-like protein 1) is an 833 amino acid protein that contains one SCA7 domain. The ATXN7L1 gene is conserved in chimpanzee, canine, mouse, rat and chicken, and maps to human chromosome 7q22.3. Chromosome 7 is about 158 million bases long, encodes over 1,000 genes and makes up about 5%of the human genome. Chromosome 7 has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATXN7L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATXN7L1
Gene Accession No. Q9ULK2
Gene Alias 2810423G08Rik; ataxin 7 like 1; ataxin 7-like 1; ataxin 7-like 4; ataxin-7-like protein 1; Ataxin-7-like protein 4; Atxn7l1; Atxn7l4; KIAA1218; RGD1305730
Gene Symbols ATXN7L1
Host Species Mouse
Immunogen Full length human recombinant protein of ATXN7L1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222255
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.