Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATXN7L2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ATXN7L2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ATXN7L2 Polyclonal specifically detects ATXN7L2 in Human samples. It is validated for Western Blot.Specifications
ATXN7L2 | |
Polyclonal | |
Rabbit | |
ataxin 7-like 2, FLJ00381, MGC46534 | |
ATXN7L2 | |
IgG | |
77 kDa |
Western Blot | |
Unconjugated | |
RUO | |
127002 | |
Synthetic peptides corresponding to ATXN7L2(ataxin 7-like 2) The peptide sequence was selected from the middle region of ATXN7L2. Peptide sequence NTPSPSFSKLPPSKASKSSKGKDGVEVEAPSRKRKLSPGPTTLKRTCILE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title