Learn More
Invitrogen™ ATXN7L3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ATXN7L3 protein.
Specifications
Specifications
| Antigen | ATXN7L3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ATXN7L3 |
| Gene Accession No. | Q14CW9 |
| Gene Alias | AT7L3; ataxin 7 like 3; ataxin 7-like 3; ataxin-7-like protein 3; ATXN7L3; DKFZp761G2113; E030022H21Rik; SAGA-associated factor 11 homolog |
| Gene Symbols | ATXN7L3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.