Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATXN7L3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584638
Description
Immunogen sequence: LRSLLTTQCG VISEHTKKMC TRSLRCPQHT DEQRRTVRIY FLGPSAVLPE VESSLDNDSF DMTDSQALIS RL.
Component of the transcription regulatory histone acetylation (HAT) complex SAGA, a multiprotein complex that activates transcription by remodeling chromatin and mediating histone acetylation and deubiquitination. Within the SAGA complex, participates in a subcomplex that specifically deubiquitinates both histones H2A and H2B (PubMed:18206972, PubMed:21746879). The SAGA complex is recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation. Within the complex, it is required to recruit USP22 and ENY2 into the SAGA complex (PubMed:18206972). Regulates H2B monoubiquitination (H2Bub1) levels. Affects subcellular distribution of ENY2, USP22 and ATXN7L3B (PubMed:27601583). [UniProt].
Specifications
ATXN7L3 | |
Polyclonal | |
Unconjugated | |
ATXN7L3 | |
AT7L3; ataxin 7 like 3; ataxin 7-like 3; ataxin-7-like protein 3; ATXN7L3; DKFZp761G2113; E030022H21Rik; SAGA-associated factor 11 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14CW9 | |
ATXN7L3 | |
Recombinant protein corresponding to Human ATXN7L3. Recombinant protein control fragment (Product #RP-107329). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction