Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATXN7L3B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989318
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9318 Supplier Bioss Supplier No. BS9754R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

By binding to ENY2, interferes with the nuclear functions of the deubiquitinase (DUB) module of the SAGA complex which consists of ENY2, ATXN7, ATXN7L3 and the histone deubiquitinating component USP22. Affects USP22 DUB activity toward histones indirectly by changing the subcellular distribution of ENY2 and altering ENY2 availability for ATXN7L3 interaction. Regulates H2B monoubiquitination (H2Bub1) levels through cytoplasmic sequestration of ENY2 resulting in loss of nuclear ENY2-ATXN7L3 association which destabilizes ATXN7L3. Affects protein expression levels of ENY2 and ATXN7L3.

Specifications

Antigen ATXN7L3B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATXN7L3B
Gene Accession No. Q96GX2
Gene Alias ataxin 7 like 3B; ataxin 7-like 3B; Ataxin-7-like protein 3B; ATXN7L3B; lnc-SCA7; putative ataxin-7-like protein 3B
Gene Symbols ATXN7L3B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ATXN7L3B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 552889
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.