Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atypical Chemokine Receptor D6 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206006
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6006 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6006 Supplier Fabgennix Inc Supplier No. ACRD6601AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Atypical Chemokine Receptor D6 Polyclonal antibody specifically detects Atypical Chemokine Receptor D6 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Atypical Chemokine Receptor D6
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ACKR2
Gene Accession No. O00590, O08707, O09027
Gene Alias Ackr2; AI464239; Atypical chemokine receptor 2; C-C chemokine receptor D6; Ccbp2; CC-chemokine-binding receptor JAB61; CCR10; CCR10-related receptor; CCR9; chemokine (C-C motif) receptor 9; chemokine (C-C) receptor 9; chemokine binding protein 2; chemokine decoy receptor D6; chemokine receptor CCR-10; Chemokine receptor CCR-9; chemokine receptor D6; chemokine-binding protein 2; Chemokine-binding protein D6; CMKBR9; D6; D6 beta-chemokine receptor; hD6
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 200-300 on ACRD6 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1238, 140473, 59289
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.