Learn More
Invitrogen™ AU1 tag Polyclonal Antibody

Description
This product is predicted to work in ELISA at a dilution of 1:10,000. The amino acid sequence for this product is: DTYRYI.
Specifications
Specifications
| Antigen | AU1 tag |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene Alias | AU1 Epitope Tag |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide DTYRYI (KLH-coupled). |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.