Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AU5 Epitope Tag Antibody, DyLight 594, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB600461DL594
Description
AU5 Epitope Tag Polyclonal antibody specifically detects AU5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
AU5 Epitope Tag | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
AU5 epitope tag, TDFYLK epitope tag, TDFYLK tag | |
Goats were immunized with AU5 (TDFYLK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
0.1 mL | |
Epitope Tags | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
DyLight 594 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Purified | |
Epitope Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction