Learn More
Beckman Coulter AU5821 Clinical Chemistry Analyzer
Shop All Beckman Coulter Products
Click to view available options
Throughput:
2,000 Tests/hr.
Description
Give your high- and ultra high-volume core laboratory a performance boost. Clinical chemistry analyzers in the AU5800 series are the fastest in the industry, processing up to 2,000 tests per hour with a full menu of assays, including Therapeutic Drug Monitoring (TDM) and Drugs of Abuse. Increase lab productivity with continuous operation, powerful throughput and scalable capacity up to four modules.
- Continuous operation with minimal downtime and easy maintenance
- Complete replacement of sample and reagent probes, mixer bars, ISE and syringes in no more than three steps, no longer than 60 seconds
- Perform routine maintenance easily, minimizing startup time
- Scalable capacity with a simplified and efficient workflow
- Increase testing capacity as needed with the modular design of the AU5800
- Connect up to four analytical modules depending on your volume requirements
- Simplify technologists' tasks with intuitive and easy-to-perform daily management procedures
- Powerful throughput with reduced analytical turnaround time
- Maximize throughput with an intelligent sample management system that optimizes the processing of racks based on the tests ordered
- Ensure quick turnaround time for those critical patients with STAT priority testing and auto-repeat of abnormal results
- Configure your total laboratory solution with options to integrate AU5800 with pre-analytical automation, immunoassay and clinical IT
Specifications
Specifications
| Type | Clinical Chemistry Analyzer |
| For Use With (Equipment) | Beckman Coulter Analyzers |
| Throughput | 2,000 Tests/hr. |
| ISE Type | Single ISE |