Learn More
Beckman Coulter AU5811 Clinical Chemistry Analyzer
Clinical chemistry analyzers in the AU5800 series are the fastest in the industry, processing up to 2,000 tests per hour with a full menu of assays.
$307438.00 - $313943.00
Specifications
For Use With (Equipment) Beckman Coulter Analyzers
Throughput 2,000 Tests/hr.
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
21-403-561
View Documents
Beckman Coulter
B04078
Single ISE
Each for $307,438.00
Only null left
Add to Cart
21-403-562
View Documents
Beckman Coulter
B04079
Dual ISE
Each for $313,943.00
Only null left
Add to Cart
Description
Description
  • Continuous operation with minimal downtime and easy maintenance
  • Complete replacement of sample and reagent probes, mixer bars, ISE and syringes in no more than three steps, no longer than 60 seconds
  • Perform routine maintenance easily, minimizing startup time
  • Scalable capacity with a simplified and efficient workflow
  • Increase testing capacity as needed with the modular design of the AU5800
  • Connect up to four analytical modules depending on your volume requirements
  • Simplify technologists' tasks with intuitive and easy-to-perform daily management procedures
  • Powerful throughput with reduced analytical turnaround time
  • Maximize throughput with an intelligent sample management system that optimizes the processing of racks based on the tests ordered
  • Ensure quick turnaround time for those critical patients with STAT priority testing and auto-repeat of abnormal results
  • Configure your total laboratory solution with options to integrate AU5800 with pre-analytical automation, immunoassay and clinical IT
Specifications
Specifications
Beckman Coulter Analyzers
2,000 Tests/hr.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.