Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aucubin, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187684 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 mg
25 mg

Aucubin is a chromogene iridoid glucoside isolated from the genus, Veronica L. and exhibits a broad-spectrum of biological activities such as hepatoprotective, antitoxic, neuroprotective, antiaging, antiosteoporosis, anti-inflammatory and cardioprotective effects.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 479-98-1
Physical Form Neat
Linear Formula C15H22O9
MDL Number MFCD00136026
Synonym Aucuboside; Rhimantin
Type Analytical Standard
Quantity 5 mg
Formula Weight 346.33
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.