Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aucubin, ≥99.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 mg
25 mg
Description
Food and Beverages
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 479-98-1 |
| Physical Form | Neat |
| Linear Formula | C15H22O9 |
| MDL Number | MFCD00136026 |
| Synonym | Aucuboside; Rhimantin |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 346.33 |
| Percent Purity | ≥99.0% (HPLC) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction