Learn More
Audit™ MicroControls™ Control FD Urine Chemistry
Click to view available options
Type:
Control FD Urine Chemistry, Level 1
Control FD Urine Chemistry, Level 2
Description
- Analytes within this kit compose Set 1 and Set 2
- One level
- Freeze Dried
- 10 x 3mL
Analytes: Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Osmolality, Phosphorus, Potassium, Sodium, Urea Nitrogen and Uric Acid
Specifications
Specifications
Type | Control FD Urine Chemistry, Level 1 |