Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AUH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546701 Supplier Invitrogen™ Supplier No. PA546701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KLAINQGMEV DLVTGLAIEE ACYAQTIPTK DRLEGLLAFK EKRPPRYKGE.

The methylglutaconyl-CoA hydratase, mitochondrial protein binds to the AU-rich element (ARE), a common element found in the 3' UTR of rapidly decaying mRNA such as c-fos, c-myc and granulocyte/ macrophage colony stimulating factor. ARE elements are involved in directing RNA to rapid degradation and deadenylation. AUH is also homologous to enol-CoA hydratase, an enzyme involved in fatty acid degradation, and has been shown to have intrinsic hydratase enzymatic activity. AUH is thus a bifunctional chimera between RNA binding and metabolic enzyme activity. A possible subcellular localization in the mitochondria has been demonstrated for the mouse homolog of this protein which shares 92% identity with the human protein. It has been suggested that AUH may have a novel role as a mitochondrial located AU-binding protein. Human AUH is expressed as a single mRNA species of 1.8 kb, and translated as a 40- kDa precursor protein which is subsequently processed to a 32- kDa mature form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AUH
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AUH
Gene Accession No. Q13825
Gene Alias 3-methylglutaconyl-CoA hydratase; AU RNA binding methylglutaconyl-CoA hydratase; AU RNA binding protein/enoyl-CoA hydratase; AU RNA binding protein/enoyl-coenzyme A hydratase; AU RNA-binding protein/enoyl-Coenzyme A hydratase; AU-binding enoyl-CoA hydratase; AU-binding protein/enoyl-CoA hydratase; Auh; AU-specific RNA-binding enoyl-CoA hydratase; C77140; Itaconyl-CoA hydratase; Methylglutaconyl-CoA hydratase, mitochondrial; muAUH; W91705
Gene Symbols AUH
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human AUH (aa 290-339).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 549
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.