Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AUNIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555964 Supplier Invitrogen™ Supplier No. PA555964
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CGVWLDAAAL KRRKVQTHLI KPGTKMLTLL PGERKANIYF TQRRAPSTGI HQRSIASFFT LQPGKTNGSD QTSVSSHTES QIN Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 67%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AUNIP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AUNIP
Gene Accession No. Q9H7T9
Gene Alias 2610002D18Rik; AIBP; Aunip; aurora A-binding; aurora A-binding protein; aurora kinase A and ninein interacting protein; aurora kinase A and ninein-interacting protein; C1orf135; clathrin heavy chain linker domain containing 1; Clhc1
Gene Symbols AUNIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AUNIP. Recombinant protein control fragment (Product #RP-94181).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79000
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.