Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AUP1 Polyclonal Antibody

Catalog No. PIPA5122221
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122221 20 μL
PIPA5118947 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122221 Supplier Invitrogen™ Supplier No. PA5122221
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AUP1 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

AUP1 contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain they may be involved in binding ubiquitin-conjugating enzymes. The unprocessed precursor is of 476 amino acids in length and has an estimated molecular weight of 53 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AUP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene AUP1
Gene Accession No. Q9Y679
Gene Alias AA589454; ancient ubiquitous protein 1; ancient ubiquitous protein 1 isoform 2; ancient ubiquitous protein 1; LOW QUALITY PROTEIN: ancient ubiquitous protein 1; Aup1; AUP1 lipid droplet regulating VLDL assembly factor; AUP1, lipid droplet regulating VLDL assembly factor; HUBCEP52; LOW QUALITY PROTEIN: ancient ubiquitous protein 1; RPL40
Gene Symbols AUP1
Host Species Rabbit
Immunogen Recombinant protein Ancient Ubiquitous Protein 1. The antigen corresponds to amino acid range 201-410 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100518187, 550
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.