Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563763
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3763 Supplier Bethyl Laboratories, Inc Supplier No. A302899A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

AUP1 contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain thay may be involved in binding ubiquitin-conjugating enzymes. The unprocessed precusor is of 476 amino acids in length and has an estimated molecular weight of 53 kDa.

Specifications

Antigen AUP1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene AUP1
Gene Accession No. Q9Y679
Gene Alias 60S ribosomal protein L40, HUBCEP52, RPL40, Ubiquitin 52 amino acid fusion protein, Ubiquitin A52 residue ribosomal protein fusion product 1, Ubiquiti
Gene Symbols AUP1
Host Species Rabbit
Immunogen Between 426 and 476
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 550
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.