Learn More
3M™ Particulate Filter 2091, P100
Shop All 3M Products
Click to view available options
:
Particulate Filter; Certifications/Compliance: NIOSH; Model: 2091; Filter Type: P100; Filter Efficiency: 99.97%; Quantity: 100/Cs.
Description
The 3M™ Particulate Filter 2091, P100 helps provide respiratory protection against exposure to lead, asbestos, cadmium, arsenic, and MDA. This filter helps protect against a wide range of oil and non-oil based particulate contaminants.
- Breathing hazardous particles can pose a risk to your health
- This filter provides at least 99.97% filter efficiency against certain oil and non-oil based particles
- Passes NIOSH (National Institute for Occupational Safety and Health) P100-series test criteria
- Assigned filter color code “magenta” in the NIOSH system
- Effective for concentrations up to 10 times the Permissible Exposure Limit (PEL) with half facepieces, or 50 times PEL with quantitatively fit-tested full facepieces
- Helps protect against a wide range of oil and non-oil based particulate contaminants
- 3M’s Advanced Electrostatic Media (AEM) provides a comfortable, lightweight, easy-breathing combination
- Commonly used for welding, brazing, torch cutting, metal pouring and soldering
- Works for many different applications, reducing inventory needs and training requirements of safety equipment
- Flame and water resistant
- Commonly used in industries including agriculture, construction, general manufacturing, pharmaceuticals, and primary metals
- Filter is directly compatible with the following equipment:
- 3M™ Half and Full Facepieces 6000, 7000 and FF-400 Series
- 3M™ Respirator 5000 Series or 3M™ Cartridge 6000 Series (If used with 3M™ Adapter 502)
- 3M™ Scott™ AV-3000 HT and AV-3000 SureSeal Facepieces (If used with the 3M™ Scott™ AV-632 Bayonet Adapter)
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Filter Type | P100 |
| Product Type | Particulate Filter |
| Filter Efficiency | 99.97% |
| For Use With (Application) | Assembly and Mechanical, Brazing, Chemical Manufacturing, Cleaning, Composite Finishing, Final Finish, Manufacturing, Metal Pouring, Paint Preparation, Painting, Pesticide Application, Petrochemical, Powder Charging, Powder Coating, Pulp and Paper, Remediation, Sanding, Soil Preparation, Soldering, Spray Coating, Torch Cutting, Welding, Wood Processing |
| Model | 2091 |
| Quantity | 100/Cs. |