Learn More
Mystaire™ Aura™ Safety Filter
For Aura™ Series Ductless Fume Hoods. Designed to incorporate both gas phase and mechanical phase filtration into one safety filter.
Supplier: Mystaire™ AU5704
Description
- Dual purpose safety filter is critical to ensuring operator and laboratory safety as it provides adequate reserve gas phase capacity as the primary filter(s) reach saturation.
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected.
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR).
Specifications
| Secondary | |
| Filter | |
| Hydrocarbons |
| Aura 30, 54, 750 Fume Hood | |
| Gas Phase Carbon | |
| Safety Filter |