Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurintricarboxylic acid, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
100g
4431-00-9
C22H14O9
422.345
MFCD00011663
GIXWDMTZECRIJT-UHFFFAOYSA-N
aurintricarboxylic acid, aluminon free acid, aluminon, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dienylidenemethylene di salicylic acid, benzoic acid, 5-3-carboxy-4-hydroxyphenyl 3-carboxy-4-oxo-2,5-cyclohexadien-1-ylidene methyl-2-hydroxy, 1,4-cyclohexadiene-1-carboxylic acid, 3-bis 3-carboxy-4-hydroxyphenyl methylene-6-oxo, 3,3'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 6-hydroxybenzoic acid, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 2-hydroxybenzoic acid
2259
CHEBI:87397
5-[(3-carboxy-4-hydroxyphenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-hydroxybenzoic acid
C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)O)C(=O)O)C(=O)O)O
4431-00-9
C22H14O9
422.345
MFCD00011663
GIXWDMTZECRIJT-UHFFFAOYSA-N
aurintricarboxylic acid, aluminon free acid, aluminon, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dienylidenemethylene di salicylic acid, benzoic acid, 5-3-carboxy-4-hydroxyphenyl 3-carboxy-4-oxo-2,5-cyclohexadien-1-ylidene methyl-2-hydroxy, 1,4-cyclohexadiene-1-carboxylic acid, 3-bis 3-carboxy-4-hydroxyphenyl methylene-6-oxo, 3,3'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 6-hydroxybenzoic acid, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 2-hydroxybenzoic acid
2259
CHEBI:87397
5-[(3-carboxy-4-hydroxyphenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-hydroxybenzoic acid
C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)O)C(=O)O)C(=O)O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Melting Point >300°C
Beilstein 2228904
Quantity 5g
Solubility Information Insoluble in water.
Formula Weight 422.35
Chemical Name or Material Aurintricarboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-628-1

RTECSNumber : GU4790000

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only