Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AURKA (Aurora A), His Tag Recombinant Protein

Catalog No. p-7075638
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length Assay Entry: Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic System Compatibility: LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate

AURKA (Aurora A) is a serine/threonine kinase that regulates mitosis by association in vivo with centrosomes. Overexpression of AURKA is observed in many types of cancer. Aurora A plays a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal. Aurora A is viewed as a potential target for anticancer drug treatment. Tissue specificity: Highly expressed in testis and weakly in skeletal muscle, thymus and spleen.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14965
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.05% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 4 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 6790
Molecular Weight (g/mol) 49.9 kDa
Name Human AURKA (Aurora A), His Tag
Quantity 1 mg
Regulatory Status For research use only. Not for use in diagnostic procedures
Gene Alias Aik; Airk; AIRK1; Ark1; ARK-1; AU019385; AURA; AURAK; AURKA; Aurora 2; aurora A; Aurora A Kinase; aurora family kinase 1; Aurora kinase A; aurora/IP; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; AURORA2; Aurora-A; aurora-related kinase 1; AW539821; Ayk1; Breast tumor-amplified kinase; BTAK; EC 2.7.11.1; hARK1; IAK; IAK1; Ipl1- and aurora-related kinase 1; IPL1-related kinase; MGC34538; OTTHUMP00000031341; OTTHUMP00000166072; PPP1R47; protein phosphatase 1, regulatory subunit 47; ratAurA; RP5-1167H4.6; sereine threonine kinase; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; serine/threonine-protein kinase 6; aurora kinase A; serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase Ayk1; STK15; STK6; STK7; Unknown (protein for MGC:134472)
Common Name Aurora A
KinaseFamily Aurora
Product Type AURKA (Aurora A) Recombinant Human Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Purity or Quality Grade 85% by SDS-PAGE
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.