Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AURKA Mouse anti-Human, Clone:2D8A4, Abnova™
Mouse monoclonal antibody raised against recombinant human AURKA.
Supplier: Abnova Corporation MAB17860
Description
The protein encoded by this gene is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression. A processed pseudogene of this gene has been found on chromosome 1, and an unprocessed pseudogene has been found on chromosome 10. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]Specifications
AURKA | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-400) Immunofluorescence (1:200-1000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
AURKA | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2D8A4 | |
aurora kinase A | |
In PBS (0.05% sodium azide) | |
AIK, ARK1, AURA, AURORA2, BTAK, MGC34538, STK15, STK6, STK7 | |
Recombinant protein corresponding to amino acids 268-404 of human AURKA from E. coli. | |
6790 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction