Learn More
Promega AURKA-NanoLuc Fusion Vector, 20microg
Catalog No. PRNV1041 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRNV1041 20 μg
1 options
This item is not returnable. View return policy
Catalog No. PRNV1041 Supplier Promega Supplier No. NV1041
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used as a standard cloning vector, a template for in vitro transcription or for production of circular ssDNA.
  • Standard Cloning Vectors That Can Produce ssDNA and In vitro Transcribe Sense and Antisense RNA
  • Blue/white screening for easy identification of recombinant clones
  • In vitro transcription using the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • The difference between + and − vectors is orientation of the f1 origin
Specifications
Quantity 20 μg
Storage Requirements -30°C to -10°C