Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AURKAIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556869 Supplier Invitrogen™ Supplier No. PA556869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KMNHHKYRKL VKKTRFLRRK VQEGRLRRKQ IKFEKDLRRI WLKAGLKEAP EGWQTPKIYL R Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

AURKA is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AURKAIP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AURKAIP1
Gene Accession No. Q9NWT8
Gene Alias 0610033H09Rik; 28S ribosomal protein S38, mitochondrial; Aip; AKIP; AURKA; AURKA-interacting protein; AURKAIP1; aurora kinase A interacting protein 1; aurora kinase A-interacting protein; aurora-A kinase interacting protein; Mitochondrial small ribosomal subunit protein mS38; Mrps38; MRP-S38
Gene Symbols AURKAIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AURKAIP1. Recombinant protein control fragment (Product #RP-94467).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.