Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AURKC Mouse anti-Human, Clone: OTI4F7, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500553

Description
Aurora kinases belong to a highly conserved family of mitotic serine/threonine kinases with three members identified among mammals: Aurora A, B, and C. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.Specifications
AURKC | |
Monoclonal | |
0.84 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UQB9 | |
AURKC | |
Full-length protein expressed in 293T cell transfected with human AURKC expression vector | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4F7 | |
Unconjugated | |
AURKC | |
AIE2, AIK3, ARK3, AurC, HEL-S-90, SPGF5, STK13, aurora-C | |
Mouse | |
Affinity Chromatography | |
RUO | |
6795 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction