Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora A Antibody (1H8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Aurora A |
---|---|
Clone | 1H8 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Aurora A Monoclonal antibody specifically detects Aurora A in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
Aurora A | |
ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7 | |
A synthesized peptide derived from Human Aurora A [UniProt O14965] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1H8 | |
ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, MAP Kinase Signaling, Mitotic Regulators, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6790 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title