Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora A Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100267AF700
Description
Aurora A Polyclonal antibody specifically detects Aurora A in Human samples. It is validated for Western BlotSpecifications
Aurora A | |
Polyclonal | |
Western Blot | |
AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7 | |
The immunogen recognized by this antibody maps to a region between residues 75 and 125 of human Aurora A Kinase using the numbering given in entry NP_003591.2 (GeneID 6790). | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, MAP Kinase Signaling, Mitotic Regulators, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
6790 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction