Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora A/B Antibody (3H1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250043AF532
Description
Aurora A/B Monoclonal antibody specifically detects Aurora A/B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Aurora A/B | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
3H1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
AIK;AIK2;AIM1;AIM-1;AIRK1;AIRK2;ARK1;ARK2;AURA;AurB;AURKA;AURKB;aurkb-sv1;aurkb-sv2;AYK1;BTAK;IAK1;IPL1;PPP1R47;PPP1R48;STK1;STK12;STK15;STK5;STK6;STK7 | |
Recombinant full length human Aurora A expressed in and purified from E. coli. [UniProt# O14965] | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction