Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora A+B+C Rabbit anti-Human∣Mouse, Polyclonal, Bioss
SDP

Catalog No. 501984735
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4735 Supplier Bioss Supplier No. BS2445R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aurora-B kinase (AurB) is a serine/threonine kinase which is the enzymatic core of the Chromosomal Passenger Complex (CPC). This complex functions in chromosome alignment and separation, histone modification, and cytokinesis. CPC also includes three non-enzymatic subunits known as the inner centromere protein (INCENP), survivin, and borealin, which determine the activity, localization, stability, and possibly even the substrate specificity of AurB. Two CPCs exist during mitosis: one containing all four members and another consisting of INCENP and AurB. Quaternary CPC functions during chromosome alignment and cytokinesis, whereas the INCENP-AurB complex may be responsible for modifying histone H3. AurB is overexpressed in a number of tumours, including primary breast and colon tumour samples. Its expression in tumours seems to be linked with Aurora-A kinase, suggesting a feedback mechanism between the two proteins.

Specifications

Antigen Aurora A+B+C
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AURKB
Gene Accession No. O70126, Q96GD4
Gene Alias AIE2; AIK1; Aik2; AIK3; AIM1; AIM-1; AIRK2; AIRK3; AL022959; Ark2; ARK-2; ARK3; ARK-3; AurB; Aurkb; aurkb-sv1; aurkb-sv2; AURKC; Aurora 1; Aurora 3; Aurora- and IPL1-like midbody-associated protein 1; aurora B; Aurora kinase B; aurora kinase B; serine/threonine-protein kinase 12; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora kinase C; Aurora/IPL1-related kinase 2; aurora-1; aurora-B; aurora-related kinase 2; BOS_18918; EC 2.7.11.1; IPL1; NP_004208; PPP1R48; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; serine/threonine kinase 5; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase aurora-B; STK1; STK-1; STK12; STK13; STK5
Gene Symbols AURKB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Aurora B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20877, 9212
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.